Order Entry
Japan
ContactUsLinkComponent
1,2-Dichloroethane, BAKER, J.T.Baker®
Flammable
Health
Irritant
1,2-Dichloroethane, BAKER, J.T.Baker®
Catalog # 9301-03
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Health
Irritant
1,2-Dichloroethane, BAKER, J.T.Baker®
Catalog # 9301-03
Supplier:  Avantor
CAS Number:  

Specifications

  • パックタイプ:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    1.245 - 1.255
  • Specific Gravity at 25°/25°C:
    1.245 - 1.255
  • Acidity:
    ≤10 ppm
  • Free Halogens:
    Passes Test
  • Lead (Pb):
    ≤1 mg/kg
  • Nonvolatile Residue:
    ≤0.002 %
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.03 %
  • Distilling Range(Lower):
    82 - 85 °C

Specification Test Results

About this item

 :
 :98.96 g/mol
Merck Index:13,03831
Boiling Pt:83,5 °C (1013 hPa)
Density:1,254 g/cm³ (20 °C)
Flash Pt:13 °C
Storage Temperature:Ambient
EINECS:203-458-1
UN:1184
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
CAS Number:107-06-2
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963