VWR® Screw Neck Vials, ND10
Supplier: VWR Collection
Certificates
About this item
ND10 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
- High performance caps/septa
- Good handling with gloves
- Quick assembly
ND10 screw neck vials are constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear versions with a flat bottom to ensure compatibility with glass or plastic inserts.
Associated screw caps mostly available with assembled septa are engineered to prevent sample contamination while providing consistent, secure seals. The use of closed caps allows the samples to be stored for a long time (long-term storage applications).