Order Entry
Japan
ContactUsLinkComponent
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Class A, Serialised
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Class A, Serialised
Catalog # VWRU10124-070
Supplier:  VWR Collection
undefined
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Class A, Serialised
Catalog # VWRU10124-070
Supplier:  VWR Collection
Specifications
  • Capacity:
    50 ml
  • Description:
    Individual, stopper no:13 and stopper type: glass pennyhead
  • Pk:
    1
Specifications
About this item
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
  • Borosilicate glass fabrication
  • Flasks are individually serialised
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with glass penny head stopper.
Ordering information: Flasks come with individual certificate of compliance.