Water Reag. Ph. Eur. 1095503 for chromatography
Catalog # 87960.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2