Masterflex® Microcentrifuge Tubes, Avantor®
Graduated Microcentrifuge Tubes with Snap caps.
- Centrifuge upto 20,000 RCF
- Withstand Temperature from -80ᵒC to +121ᵒC
- Rnase, Dnase, Endotoxin and Pyrogen Safe
Cole-Parmer microcentrifuge tubes are manufactured from FDA approved resin. Tubes include an attached cap. All tubes feature a frosted writing surface on the tube and are graduated. Boil-proof tubes are guaranteed to stay closed, even during boiling or autoclaving. All tubes are supplied in double safety self-standing bulk bags.
- Environmentally Preferable:
Frequently Bought Together