Order Entry
Japan
ContactUsLinkComponent
Burkle® Drum and Barrel Port Adapters
Burkle 5601-0130 Screw Joint for Drums and Barrels, SS, 2" fine thread, gastight
  MFLX06043-83
undefined
Burkle 5601-0130 Screw Joint for Drums and Barrels, SS, 2" fine thread, gastight
  MFLX06043-83
  • Description:
    Screw Joint for Drums and Barrels, SS, 2" fine thread, gastight
  • Material:
    Stainless steel
  • Warranty:
    15 Months
  • Compatible with:
    2" fine thread
  • Pk:
    1
Securely attach drum/barrel pumps.
  • Adapt your system to the Burkle drum and barrel systems
Screw joints and bungs in a variety of materials.
  • Description:
    Screw Joint for Drums and Barrels, SS, 2" fine thread, gastight
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Warranty:
    15 Months
  • Compatible with:
    2" fine thread
  • Pk:
    1