Female Threaded Cap Fittings, PTFE
Female Threaded Pipe Cap, ⅛" NPT(F), PTFE, Threaded Cap, PTFE, Straight, ⅛"
Catalog # MFLX34030-61
CE certificate
Supplier: Avantor Fluid Handling
Female Threaded Cap Fittings, PTFE
Catalog # MFLX34030-61
Supplier: Avantor Fluid Handling
CE certificate
Jump to:
Product Details & Documents
Chemically-inert PTFE handles most aggressive chemicals.
- Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.
Specifications
- Connection Type:Threaded Cap
- Flow pattern:Straight
- Thread size NPT(F) (port 1):⅛"
- Description:Female Threaded Pipe Cap, ⅛" NPT(F), PTFE
- Fitting material:PTFE
- Cat. No.:MFLX34030-61
- Warranty:2 years
- Sterilisation:Autoclave or ethylene oxide (ETO)