Order Entry
Japan
ContactUsLinkComponent
Miniature Plastic Hose Barb Check Valves
undefined
Miniature Plastic Hose Barb Check Valves
  • Int. Ø tubing:
  • 0,1875"
  • 0,125"
  • 0,25"
  • 0,25"
  • Max. pressure:
  • 50 psi
  • 50 psi
  • 125 psi
  • 50 psi
  • Max. temperature:
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 93 °C (200 °F)
  • Wetted materials:
  • Nylon and Flourosilicone
  • Nylon and Flourosilicone
  • PVDF and Viton
  • Polypropylene and Viton
  • Working pressure:
  • 5 psi
  • 5 psi
  • 5 psi
  • 5 psi
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together