Acetonitrile, ultra-low water content ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED®, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Density:0.79 g/cm³
Flash Pt:2.00 °C
UN:1648
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm:≤0.010
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤190
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Appearance:Passes Test
- Assay (CH₃CN) (by GC, corrected for water):≥99.8 %
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- Titrable Acid (µeq/g):≤0.8
- Titrable Base (µeq/g):≤0.6
- Water (by KF, coulometric):≤5 ppm
Specification Test Results
Frequently Bought Together