tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), granular USP, FCC, J.T.Baker®
Catalog # 3649-K1
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
- Pk Info:10098 g +250 g
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specification Test Results
About this item
:
:294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.70 g/cm³
Storage Temperature:Ambient
EINECS:200-675-3
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031