Order Entry
Japan
ContactUsLinkComponent
 
Sodium chloride 99-100.5%, granular, AR®, Macron Fine Chemicals™
  MACR7544-06
 :  
Sodium chloride 99-100.5%, granular, AR®, Macron Fine Chemicals™
  MACR7544-06
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

 

 

  • Assay (NaCl) (dried basis):
    99.5 - 100.5 %
  • Appearance (white, crystalline granules):
    Passes Test
  • Solubility (10 in 50):
    Passes Test
  • Identification A:
    Passes Test
  • Identification B:
    Passes Test
  • Loss on Drying at 105°C:
    ≤0.5 %
  • Acidity or Alkalinity:
    Passes Test
  • Limit of Bromide:
    ≤0.010 %
  • Calcium and Magnesium (as Ca):
    ≤0.005 %
  • Magnesium and Alkaline Earth Metals (as Ca):
    ≤0.01 %
  • Phosphate (PO₄):
    ≤0.0025 %
  • Barium (Ba):
    Passes Test
  • Arsenic (As):
    ≤0.0001 %
  • Heavy Metals (as Pb):
    ≤0.0005 %
  • Iron (Fe):
    ≤0.0002 %
  • Sulfate (SO₄):
    ≤200 ppm
  • Iodide (I):
    Passes Test
  • Copper (Cu):
    ≤0.0002 %
  • Sodium Ferrocyanide:
    Passes Test

 

 

 :
 :58.44 g/mol
 :14,08599
 :1413 °C (1013 hPa)
 :801 °C
 :2.16 g/cm³ (20 °C)
 :Management of Change (MOC) category = N
 :Ambient
 :231-598-3
 :7647-14-5
 :NaCl
 :MFCD00003477