Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR® Desiccators
Supplier:   VWR Collection
VWR® Desiccators
Supplier:  VWR Collection
Certificates
VWR® Desiccators
VWR® Desiccators
5 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass.
  • Precisely ground flat flanges and interchangeable knobbed lids
  • Base and lid with flat flange ground to fit
  • High mechanical, thermal and chemical resistance
These desiccators are ideal for drying, drying processes and protecting samples from the effects of air and moisture.
 : DIN 12491
Product Family Options
Desiccators
Product Information
  • Nominal size
    Usable height
    Ø ext.×H
    Ø int.
    Availability
    Price
  • 100 DN
    65 mm
    153×195 mm
    92 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 150 DN
    95 mm
    215×265 mm
    143 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 200 DN
    130 mm
    270×315 mm
    192 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 250 DN
    140 mm
    320×370 mm
    239 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 300 DN
    165 mm
    380×445 mm
    285 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
Spare bases for VWR desiccators
Product Information
  • Catalog #
    Usable height
    Nominal size
    Description
    Ø ext.×H
    Availability
    Price
  • 120 mm
    250 DN
    Base
    320×235 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00
    $0.00
    1 * 1 items
       $0.00
Product Information
  • Catalog #
    Usable height
    Nominal size
    Description
    Ø ext.×H
    Availability
    Price
  • 115 mm
    200 DN
    Base
    270×202 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00
    $0.00
    1 * 1 items
       $0.00
Product Information
  • Catalog #
    Usable height
    Nominal size
    Description
    Ø ext.×H
    Availability
    Price
  • 81 mm
    150 DN
    Base
    215×154 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00
    $0.00
    1 * 1 items
       $0.00
Product Information
  • Catalog #
    Usable height
    Nominal size
    Description
    Ø ext.×H
    Availability
    Price
  • 150 mm
    300 DN
    Base
    380×283 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00
    $0.00
    1 * 1 items
       $0.00
Product Information
  • Catalog #
    Usable height
    Nominal size
    Description
    Ø ext.×H
    Availability
    Price
  • 58 mm
    100 DN
    Base
    153×112 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00
    $0.00
    1 * 1 items
       $0.00
Spare lids for VWR desiccators
Product Information
  • Catalog #
    Nominal size
    Description
    Type
    Availability
    Price
  • 150 DN
    Cover
    With knob
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00
    $0.00
    1 * 1 items
       $0.00
Product Information
  • Catalog #
    Nominal size
    Description
    Type
    Availability
    Price
  • 250 DN
    Cover
    With knob
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00
    $0.00
    1 * 1 items
       $0.00
Product Information
  • Catalog #
    Nominal size
    Description
    Type
    Availability
    Price
  • 100 DN
    Cover
    With knob
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00
    $0.00
    1 * 1 items
       $0.00
Product Information
  • Catalog #
    Nominal size
    Description
    Type
    Availability
    Price
  • 200 DN
    Cover
    With knob
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00
    $0.00
    1 * 1 items
       $0.00
Product Information
  • Catalog #
    Nominal size
    Description
    Type
    Availability
    Price
  • 300 DN
    Cover
    With knob
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00
    $0.00
    1 * 1 items
       $0.00