Order Entry
Japan
ContactUsLinkComponent
VWR® 24-Hour Urine Container
VWR® 24-Hour Urine Container
Catalog # 216-2327
Supplier:  VWR Collection
undefined
VWR® 24-Hour Urine Container
Catalog # 216-2327
Supplier:  VWR Collection

Specifications

  • Capacity:
    3000 ml
  • Description:
    Urine container, brown
  • Pk:
    30

Specifications

About this item

24-hour 3 L vacuum urine container with aspiration system for vacuum tubes directly integrated in the screw cap.

  • Optimal handling: Ergonomic moulded handle
  • Graduated
  • Polyethylene

The 24 hour urine collection container is suitable for collecting urine over a timed period.

The 3 L container is brown coloured to prevent the premature decomposition of light sensitive parameters such as urobilinogen.

The container is fitted with an integrated transfer device to enable hygienic, clean sample transfer into a urine tube.

Efficiency in collecting biological samples.
New translucent vertical stripe: This solution allows operators to control the level of urine inside the container, avoiding the risk of contamination, speeding up operations and workflow in the laboratory.

Certifications: IVD

Urine straws

Product Information