Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Sealing Membranes, Classic
VWR®, Sealing Membranes, Classic
Supplier:   VWR Collection
VWR®, Sealing Membranes, Classic
Supplier:  VWR Collection
Certificates
VWR®, Sealing Membranes, Classic
VWR®, Sealing Membranes, Classic
3 Options Available Below
Product Details & Documents
VWR® PCR membranes are DNAse- and RNAse-free.
  • Minimises sample loss due to vaporisation
  • Eliminates chances of leaching
Preferred solution for sealing.
Product Family Options
Product Information
  • Availability
    Price
  • VWRA734-3330
    PCR 96-well plate sealing membrane
    PCR 96-well plate sealing membrane
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $77.84
    $77.84
    1 * 100 items
       $77.84
  • VWRA734-3331
    qPCR 96-well plate optical sealing, pressure sensitive membrane
    qPCR 96-well plate optical sealing, pressure sensitive membrane
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $394.39
    $394.39
    1 * 100 items
       $394.39
  • VWRA734-3332
    Aluminum membrane
    Aluminum membrane
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $90.54
    $90.54
    1 * 100 items
       $90.54