VWR® Maximum, Cut Resistant Gloves
About this item
This VWR® glove with polyurethane coating provides superior cut protection for applications that include a high risk of cuts.
- Nylon/spandex/polyester blend composition
- Polyurethane (PU) coating on palm and fingertips
- Grey knitted glove with grey coating
Ideal for handling items or tools with a high risk of cuts, including steel or glass handling, assembly, maintenance, and janitorial applications.
: Meets requirements for EN 388:2016 (4X42E). Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
6 Options Available Below
Product Details & Documents
This VWR® glove with polyurethane coating provides superior cut protection for applications that include a high risk of cuts.
- Nylon/spandex/polyester blend composition
- Polyurethane (PU) coating on palm and fingertips
- Grey knitted glove with grey coating
Ideal for handling items or tools with a high risk of cuts, including steel or glass handling, assembly, maintenance, and janitorial applications.
: Meets requirements for EN 388:2016 (4X42E). Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Specifications for Product Family
Specifications
- Catalog No:
- 112-5691
- 112-5692
- 112-5693
- 112-5694
- 112-5695
- 112-5696
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Finish:
- PU palm coating
- PU palm coating
- PU palm coating
- PU palm coating
- PU palm coating
- PU palm coating
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
- EN 388:2016 (4X42E)
Specifications
Product Family Options
Product Information
- Glove sizeColourAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.112-5691Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLatex / latex-freeLatex-freeFinishPU palm coatingGauge13Cuff styleKnit wristEN normEN 388:2016 (4X42E) - Specifications:
More
Specifications
Cat. No.112-5692Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLatex / latex-freeLatex-freeFinishPU palm coatingGauge13Cuff styleKnit wristEN normEN 388:2016 (4X42E) - Specifications:
More
Specifications
Cat. No.112-5693Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLatex / latex-freeLatex-freeFinishPU palm coatingGauge13Cuff styleKnit wristEN normEN 388:2016 (4X42E) - Specifications:
More
Specifications
Cat. No.112-5694Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLatex / latex-freeLatex-freeFinishPU palm coatingGauge13Cuff styleKnit wristEN normEN 388:2016 (4X42E) - Specifications:
More
Specifications
Cat. No.112-5695Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLatex / latex-freeLatex-freeFinishPU palm coatingGauge13Cuff styleKnit wristEN normEN 388:2016 (4X42E) - Specifications:
More
Specifications
Cat. No.112-5696Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLatex / latex-freeLatex-freeFinishPU palm coatingGauge13Cuff styleKnit wristEN normEN 388:2016 (4X42E)