VWR®, Pipette Controller, Classic
About this item
Pipette controller for glass and plastic pipettes from 1 to 100 ml.
- Speed of suction and dispensing controlled through the finger triggers
- Fully charged has approximately 8 h of continuous use available
- Low battery light illuminates when 1 hour's power remains
Offers a unique combination of features to give precision fingertip control in a classic streamlined design, suitable for users with large or small hands. Gravity dispense mode is designed for use with 'To Deliver' (TD) pipettes.
4 Options Available Below
Product Details & Documents
Pipette controller for glass and plastic pipettes from 1 to 100 ml.
- Speed of suction and dispensing controlled through the finger triggers
- Fully charged has approximately 8 h of continuous use available
- Low battery light illuminates when 1 hour's power remains
Offers a unique combination of features to give precision fingertip control in a classic streamlined design, suitable for users with large or small hands. Gravity dispense mode is designed for use with 'To Deliver' (TD) pipettes.
Product Family Options
Product Information
- Colour codeAvailabilityPrice