Order Entry
Japan
ContactUsLinkComponent
VWR®, Platform Shakers
VWR®, Platform Shakers
Supplier:  VWR Collection
Certificates
undefined
VWR®, Platform Shakers
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • VWRA444-1229
  • VWRA444-1230
  • Speed:
  • 50 - 500 min⁻¹
  • 50 - 500 min⁻¹
  • Shaking motion:
  • Orbital
  • Linear
  • Max. load:
  • 10 kg
  • 10 kg
  • Weight:
  • 7,5 kg
  • 7,5 kg
  • W×D×H:
  • 350×408×99 mm
  • 350×408×99 mm