Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR® PHP Plates

VWR® PHP Plates

VWR® PHP Plates
Supplier:  VWR Collection
Certificates
VWR® PHP Plates
VWR® PHP Plates

2 Options Available Below

Product Details & Documents

High performance, gamma sterilized and disposable Petri dishes for every microbiology lab.

  • Manufactured in 10K cleanroom for dust free final products
  • Sterilized as per 10⁻⁶ gamma irradiation
  • Ensures highest possible flatness for zero wastage of media (A to B and C to D lesser than 0,01 mm)
  • Made of US FDA approved PS with best-in-class clarity and tensile strength
  • Rimming of plates are designed for better stacking, marking patch for writing

Manufactured under strict aseptic conditions, VWR® Petri plates are perfect for use in automated plate filing systems. The high transparency of these plates ensures accurate observation and measurement, reducing the margin for errors in research.

Delivery information: The innovative packaging of these PHP plates is breakage proof.

Packaging: The innovative packaging of these PHP plates is breakage proof.

Product Family Options

Product Information

  • Ø
    Sterility
    Packed
    Availability
    Price
  • 391-0691
    PHP Petri plate, stack size 10
    PHP Petri plate, stack size 10
    90 mm
    Gamma irradiated
    10×60
    Log in for availability
    Log in for availability
    1 * 600 items
    1 * 600 items - $84.07
    $84.07
    1 * 600 items
       $84.07

  • 391-0693
    PHP Petri plate, stack size 20
    PHP Petri plate, stack size 20
    90 mm
    Gamma irradiated
    20×30
    Log in for availability
    Log in for availability
    1 * 600 items
    1 * 600 items - $77.84
    $77.84
    1 * 600 items
       $77.84