Order Entry
Japan
ContactUsLinkComponent
Sodium hydrogen carbonate kit 99.0-100.5% (dried basis), powder USP, FCC, J.T.Baker®
Sodium hydrogen carbonate kit 99.0-100.5% (dried basis), powder USP, FCC, J.T.Baker®
Catalog # 3509-K2
Supplier:  Avantor
CAS Number:  
Sodium hydrogen carbonate kit 99.0-100.5% (dried basis), powder USP, FCC, J.T.Baker®
Catalog # 3509-K2
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Bag
  • Pk:
    1 KIT
  • Seq:
    0075
  • Pk Info:
    1×360 g and 1×200 g

Specifications

Specification Test Results

  • Kit Components Meet Requirements:
    Passes Test

Specification Test Results

About this item

 :
 :84.01 g/mol
Merck Index:13,08655
Melting Pt:270 °C
Density:2.20 g/cm³
 :Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:205-633-8
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528