Order Entry
Japan
Orders LinkContactUsLinkComponent
 

L(+)-Histidine USP, Multi-Compendial, J.T.Baker®

Supplier:   Avantor
L(+)-Histidine USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Merck Index:13,04741
  • Melting Pt:272…273 °C
  • Density:1.44 g/cm³ (20 °C)
  •  :Management of Change (MOC) category = R
  • Storage Temperature:Ambient
  • EINECS:200-745-3
Specifications for Product Family

Specification Test Results

  • USP - Assay (C₆H₉N₃O₂) (dried basis):
    98.5 - 101.5 %
  • USP - Identification:
    Passes Test
  • USP - Specific Rotation [Å]²⁵ ^D (+):
    12.6 - 14.0 Degree
  • USP - pH:
    7.0 - 8.5
  • USP - Loss on Drying at 105°C:
    ≤ 0.2 %
  • USP - Residue on Ignition:
    ≤ 0.4 %
  • USP - Chloride (Cl):
    ≤ 0.05 %
  • USP - Sulfate (SO₄):
    ≤ 0.03 %
  • USP - Iron (Fe):
    ≤ 0.003 %
  • USP - Related Compounds - Individual Impurities:
    ≤ 0.5 %
  • USP - Related Compounds - Total Impurities:
    ≤ 2.0 %
  • EP/BP - Assay (C₆H₉N₃O₂) (dried basis):
    98.5 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Specific Rotation [Å]²º^D (+):
    11.4 - 12.4 Degree
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤ 0.2 %
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤ 0.5 %
  • EP/BP - Chloride (Cl):
    ≤ 200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤ 0.02 %
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Loss on Drying:
    ≤ 0.5 %
  • EP/BP - Ash (sulfated):
    ≤ 0.1 %
  • JP - Assay (C6H9N3O2) (dried basis):
    99.0 - 101.0 %
  • JP - Ammonium (NH₄):
    ≤ 0.02 %
  • JP - Chloride (Cl):
    ≤ 0.021 %
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤ 10 ppm
  • JP - Iron (Fe):
    ≤ 10 ppm
  • JP - Identification:
    Passes Test
  • JP - Loss on Drying at 105°C:
    ≤ 0.30 %
  • JP- Optical Rotation [Å]²º^D (+):
    11.8 - 12.8 Degree
  • JP - Related Substances:
    Passes Test
  • JP - pH:
    7.0 - 8.5
  • JP - Residue on Ignition:
    ≤ 0.10 %
  • JP - Sulfate (SO₄):
    ≤ 0.028 %
  • Bacterial endotoxins, EU/g:
    ≤ 10
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.