di-Potassium hydrogen phosphate kit USP, Multi-Compendial, J.T.Baker®
Catalog # 3250-X1
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Bag/kit
- Pk:62,869 kg
- Seq:4472
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specification Test Results
About this item
:
:174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
:Management of Change (MOC) category = HR
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383