Order Entry
Japan
ContactUsLinkComponent
 
Propionic acid ≥99% (by GC), J.T.Baker®
  U340-09
 :  Avantor
 :  
undefined
Propionic acid ≥99% (by GC), J.T.Baker®
  U340-09
 :  Avantor
 :  

 

  • Pk:
    430 lb
  • Seq:
    7293
  • Pk Info:
    1×195,04 L

 

 

  • Assay (by GC) (CH₃CH₂COOH):
    ≥99 %

 

 

 :
 :74.08 g/mol
 :13,07917
 :141 °C (1013 hPa)
 :–21 °C
 :0.99 g/cm³ (20 °C)
 :49 °C
 :Ambient
 :201-176-3
 :3463
 :8 (3),II
 :Management of Change (MOC) category = R
 :01-2119486971-24
 :GHS02
 :GHS05
 :GHS06
 :79-09-4
 :C₃H₆O₂
 :MFCD00002756