Polyethylene glycol 1,450, waxy soft solid, J.T.Baker®
U220-07
: Avantor
:
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 mL
- Seq:2725
- Pk Info:1×500 ml
- Appearance:Passes Test
- Identification (by IR):Passes Test
:
:1450 g/mol
:Management of Change (MOC) category = N
:Ambient
:500-038-2
:25322-68-3
:H(OCH₂CH₂)nOH
:MFCD00081839