Order Entry
Japan
ContactUsLinkComponent
Phenol ≥89.0% liquefied (std of Merck) USP, Multi-Compendial, J.T.Baker®
Health
Toxic
Corrosive
Phenol ≥89.0% liquefied (std of Merck) USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Health
Toxic
Corrosive
Phenol ≥89.0% liquefied (std of Merck) USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • USP - Distilling Range - Final Temperature:
    ≤182.5 °C
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Clarity of Solution and Reaction:
    Passes Test
  • USP - Nonvolatile Residue:
    ≤0.05 %
  • USP - Assay:
    ≥89.0 %
  • USP - Benzene:
    ≤2.0 ppm
  • JP - Identification (1):
    Passes Test
  • JP - Identification (2):
    Passes Test
  • JP - Boiling Point:
    ≤182 °C
  • JP - Clarity and Color of Solution and Acidity or Alkalinity:
    Passes Test
  • JP - Residue on Evaporation:
    ≤0.05 %
  • JP - Assay:
    ≥88.0 %
  • Assay (Merck):
    89.0 - 91.5 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together