Order Entry
Japan
ContactUsLinkComponent
Glycerine (vegetable based) 100% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Glycerine (vegetable based) 100% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 5093-19
Supplier:  Avantor
CAS Number:  
undefined
Glycerine (vegetable based) 100% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 5093-19
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    19 L
  • Seq:
    5282
  • Pk Info:
    1×19 L

Specifications

Specification Test Results

  • ACS - Assay (C₃H₅(OH)₃) (by GC, corrected for water):
    ≥99.5 %
  • ACS - Acrolein and Glucose:
    Passes Test
  • ACS - Chlorinated Compounds (as Cl):
    ≤0.003 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Fatty Acid Esters (as Butyric Acid):
    ≤0.05 %
  • ACS - Heavy Metals (as Pb):
    ≤2 ppm
  • ACS - Neutrality:
    Passes Test
  • ACS - Residue after Ignition:
    ≤0.005 %
  • ACS - Substances Darkened by H₂SO₄:
    Passes Test
  • ACS - Sulfate (SO₄):
    ≤0.001 %
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.5 %

Specification Test Results

About this item

 :
 :92.09 g/mol
 :Management of Change (MOC) category = N
Flash Pt:18.17 °C
Storage Temperature:Ambient
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722