


Dichloromethane : Acetone mixture 50 : 50 (v/v), ULTRA RESI-ANALYZED®, J.T.Baker®
Supplier: Avantor
Certificates
About this item
:
ADR:3, 6.1,II
:Management of Change (MOC) category = NI
Flash Pt:–20 °C
UN:1992
:GHS02
:GHS07
:GHS08
Specification Test Results
- Acetone - Assay (by GC, corrected for water):≥99.4 %
- Acetone - Color (APHA):≤10
- Acetone - Residue after Evaporation:≤1 ppm
- Acetone - Substances Reducing Permanganate:Passes Test
- Acetone - Titrable Acid (µeq/g):≤0.3
- Acetone - Titrable Base (µeq/g):≤0.60
- Water (H₂O)(by Karl Fischer titrn):≤0.5 %
- Acetone - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- Acetone - GC-ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (ng/L):≤10
- Methylene Chloride - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- Methylene Chloride - GC-ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (ng/L):≤10
- Methylene Chloride - Assay (by GC, corrected for water, exclusive of preservative):≥99.8 %
- Methylene Chloride - Color (APHA):≤10
- Methylene Chloride - Residue after Evaporation:≤1.0 ppm
- Methylene Chloride - Titrable Acid (µeq/g):≤0.30
- Methylene Chloride - Chloride (Cl):≤10 ppm
- Methylene Chloride - Water (by KF, coulometric):≤0.02 %
Specification Test Results
Frequently Bought Together