Magnesium sulphate, anhydrous 99.0-100.5% (excluding ignited substances), powder USP, Macron Fine Chemicals™
Catalog # MACR5054-26
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:200 lb
- Seq:7197
- Pk Info:1×90,71 kg
Specifications
Specification Test Results
- Appearance:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Loss on Drying at 105°C:≤ 2 %
- Loss on Ignition:≤ 2.0 %
- USP - pH (1 in 20):5.0 - 9.2
- USP - Chloride (Cl):≤ 0.014 %
- USP - Selenium (Se):≤ 0.003 %
- USP - Iron (Fe):≤ 20 ppm
- USP - Assay (as MgSO4) (ignited basis):99.0 - 100.5 %
- Solubility:Passes Test
- Thru U.S. No. 100 Sieve:≥ 85 %
Specification Test Results
About this item
:
:120.37 g/mol
Merck Index:12,05731
Melting Pt:1127 °C
:Management of Change (MOC) category = R
Storage Temperature:Ambient
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110