Order Entry
Japan
ContactUsLinkComponent
Sodium hydroxide ≥97% (by acidimetry) low in mercury, J.T.Baker®
Corrosive
Sodium hydroxide ≥97% (by acidimetry) low in mercury, J.T.Baker®
Catalog # 5045-07
Supplier:  Avantor
CAS Number:  
Corrosive
Sodium hydroxide ≥97% (by acidimetry) low in mercury, J.T.Baker®
Catalog # 5045-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    25 lb
  • Seq:
    7023
  • Pk Info:
    1×11,33 kg

Specifications

Specification Test Results

  • Assay (NaOH) (by acidimetry):
    ≥97 %
  • Sodium Carbonate (Na₂CO₃):
    ≤1.0 %
  • Chloride (Cl):
    ≤0.005 %
  • Nitrogen Compounds (as N):
    ≤0.001 %
  • Phosphate (PO₄):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.003 %
  • Ammonium Hydroxide Precipitate:
    ≤0.02 %
  • Heavy Metals (as Ag):
    ≤0.002 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Nickel (Ni):
    ≤0.001 %
  • Potassium (K)(by FES):
    ≤0.02 %
  • Trace Impurities - Mercury (Hg):
    ≤0.010 ppm

Specification Test Results

About this item

 :
 :40 g/mol
ADR:8,II
 :Management of Change (MOC) category = N
UN:1823
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548