Disodium hydrogen phosphate heptahydrate 98.0-100.5% (dry basis) USP, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7782-85-6
:
Formula:HNa₂O₄P·7H₂O
MDL Number:MFCD00149180
:268.07 g/mol
:Management of Change (MOC) category = HR
Specification Test Results
- Appearance (Colorless or translucent crystals or granules):Passes Test
- USP - Assay (Na₂HPO₄) (dried basis):98.0 - 100.5 %
- USP - Arsenic (As):≤ 16 ppm
- USP - Chloride (Cl):≤ 0.06 %
- USP - Endotoxin Concentration (EU/g):≤ 2.5
- USP - Identification:Passes Test
- USP - Insoluble Substances:≤ 0.4 %
- USP - Loss on Drying:43.0 - 50.0 %
- USP - Sulfate (SO₄):≤ 0.2 %
- EP - Identification A:Passes Test
- EP - Identification C:Passes Test
- EP - Identification D:Passes Test
- EP - Reducing Substances:Passes Test
- EP - Monosodium Phosphate:≤ 2.5 %
- EP - Chloride (Cl):≤ 200 ppm
- EP - Sulfate (SO₄):≤ 500 ppm
- EP - Appearance of Solution:Passes Test
- EP - Iron (Fe):≤ 20 ppm
- EP - Assay (anhydrous basis):98.0 - 101.0 %
- JP - Arsenic (As):≤ 2 ppm
- JP - Assay (Na₂HPO₄) (dried basis):98.0 - 101.0 %
- JP - Carbonate (CO₃):Passes Test
- JP - Chloride (Cl):≤ 0.014 %
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals:≤ 10 ppm
- JP - Identification:Passes Test
- JP - pH (1 in 50):9.0 - 9.4
- JP - Sulfate (SO₄):≤ 0.038 %
Specification Test Results
Frequently Bought Together