Sodium hydrogen carbonate 99.0-100.5% (dry basis), powder USP, FCC, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
:Management of Change (MOC) category = R
Specification Test Results
- USP - Identification:Passes Test
- USP - Limit of Sulfur Compounds:≤0.015 %
- USP - Loss on Drying:≤0.25 %
- USP - Insoluble Substances:Passes Test
- USP - Normal Carbonate:Passes Test
- USP - Limit of Ammonia:≤20 ppm
- USP - Chloride (Cl):≤0.015 %
- USP - Arsenic (As):≤2 ppm
- USP - Assay (NaHCO₃) (dried basis):99.0 - 100.5 %
- FCC - Identification:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Loss on Drying:≤0.25 %
- FCC - Assay (NaHCO₃) (dried basis):99.0 - 100.5 %
- Appearance (White powder):Passes Test
Specification Test Results
Frequently Bought Together