Potassium acetate 99.0-100.5% (dried basis), crystals USP, Macron Fine Chemicals™
MACR6696-16
:
- Pk:500 g
- Seq:3645
- Pk Info:1×500 g
- USP - Assay (CH₃COOK) (dried basis):99.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Loss on Drying at 150°C:≤1.0 %
- USP - pH of 5% Aqueous Solution:7.5 - 8.5
- USP - Sodium (Na):≤0.03 %
- Solubility:Passes Test
- USP - Heavy Metals (as Pb):≤0.002 %
:
:98.14 g/mol
:13,07687
:304 °C
:1.57 g/cm³ (25 °C)
:Management of Change (MOC) category = R
:>250 °C
:Ambient
:204-822-2
:127-08-2
:CH₃COOK
:MFCD00012458