Order Entry
Japan
ContactUsLinkComponent
Chloroform ≥99.8% (by GC, corrected for water content), purified, J.T.Baker®
Irritant
Health
Chloroform ≥99.8% (by GC, corrected for water content), purified, J.T.Baker®
Catalog # 9185-WJ
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Health
Chloroform ≥99.8% (by GC, corrected for water content), purified, J.T.Baker®
Catalog # 9185-WJ
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1.000 L
  • Seq:
    5037
  • Pk Info:
    1×1000 L

Specifications

Specification Test Results

  • Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water):
    ≥99.8 %
  • Color (APHA):
    ≤5
  • Specific Gravity at 25°/25°C:
    1.476 - 1.480
  • Nonvolatile Residue (w/v):
    ≤0.002 %
  • Free Chlorine:
    Passes Test
  • Readily Carbonizable Substances:
    Passes Test
  • Chlorinated Decomposition Products and Chloride:
    Passes Test
  • Acid and Phosgene:
    Passes Test
  • Aldehydes and Ketones (as HCHO):
    Passes Test

Specification Test Results

About this item

 :
 :119.38 g/mol
Melting Pt:–63 °C
ADR:6.1,III
 :Management of Change (MOC) category = NI
UN:1888
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826