Order Entry
Japan
ContactUsLinkComponent
 
 
 
 
Methanol ≥99.8% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
  9067-35
 :  Avantor
 :  
undefined
 
 
 
Methanol ≥99.8% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
  9067-35
 :  Avantor
 :  

 

  • Pk:
    1.625 lb
  • Seq:
    7374
  • Pk Info:
    1×737,08 kg

 

 

  • Assay (CH₃OH) (by GC, corrected for water):
    ≥ 99.8 %
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Substances Reducing Permanganate:
    Passes Test
  • Solubility in H₂O:
    Passes Test
  • Color (APHA):
    ≤ 10
  • Water (H₂O):
    ≤ 0.1 %
  • Residue after Evaporation:
    ≤ 0.001 %
  • Carbonyl Compounds - Acetaldehyde:
    ≤ 0.001 %
  • Carbonyl Compounds - Acetone:
    ≤ 0.001 %
  • Carbonyl Compounds - Formaldehyde:
    ≤ 0.001 %
  • Titrable Acid (µeq/g):
    ≤ 0.3
  • Titrable Base (µeq/g):
    ≤ 0.2
  • Trace Impurities - Copper (Cu):
    ≤ 0.10 ppm
  • Trace Impurities - Heavy Metals:
    ≤ 0.5 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 0.1 ppm
  • Trace Impurities - Nickel (Ni):
    ≤ 0.1 ppm
  • curve, no extraneous peaks):
    ≤ 0.01
  • Ultraviolet Absorbance(1.00-cm cell vs. water) - 280-400 nm (smooth:
    ≤ 0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:
    ≤ 0.040
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:
    ≤ 0.100
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:
    ≤ 0.200
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤ 0.400
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:
    ≤ 0.80
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 205 nm:
    ≤ 1.00

 

 

 :
 :32.04 g/mol
 :3,II
 :Management of Change (MOC) category = N
 :1230
 :GHS02
 :GHS06
 :GHS08
 :67-56-1
 :CH₃OH
 :MFCD00004595