


Nitric acid 69,0 - 70,0%, Finyte®, J.T.Baker®
Catalog # 5856-IN
Supplier: Avantor
CAS Number:
Specifications
- Pk:605 lb
- Seq:7353
- Pk Info:1×274,42 L
Specifications
Specification Test Results
- Assay (HNO₃):69.0 - 70.0 %
- Appearance:Passes Test
- Color (APHA):≤5
- Chloride (Cl):≤0.05 ppm
- Phosphate (PO₄):≤0.05 ppm
- Sulfate (SO₄):≤0.2 ppm
- Trace Impurities - Aluminum (Al):≤5.0 ppb
- Arsenic and Antimony (as As):≤3.0 ppb
- Trace Impurities - Barium (Ba):≤5.0 ppb
- Trace Impurities - Beryllium (Be):≤5.0 ppb
- Trace Impurities - Bismuth (Bi):≤5.0 ppb
- Trace Impurities - Boron (B):≤5.0 ppb
- Trace Impurities - Cadmium (Cd):≤5.0 ppb
- Trace Impurities - Calcium (Ca):≤10.0 ppb
- Trace Impurities - Chromium (Cr):≤10.0 ppb
- Trace Impurities - Cobalt (Co):≤5.0 ppb
- Trace Impurities - Copper (Cu):≤5.0 ppb
- Trace Impurities - Gallium (Ga):≤5.0 ppb
- Trace Impurities - Germanium (Ge):≤5.0 ppb
- Trace Impurities - Gold (Au):≤5.0 ppb
- Trace Impurities - Iron (Fe):≤5.0 ppb
- Trace Impurities - Lead (Pb):≤5.0 ppb
- Trace Impurities - Lithium (Li):≤5.0 ppb
- Trace Impurities - Magnesium (Mg):≤5.0 ppb
- Trace Impurities - Manganese (Mn):≤5.0 ppb
- Trace Impurities - Molybdenum (Mo):≤5.0 ppb
- Trace Impurities - Nickel (Ni):≤5.0 ppb
- Trace Impurities - Niobium (Nb):≤5.0 ppb
- Trace Impurities - Potassium (K):≤5.0 ppb
- Trace Impurities - Silicon (Si):≤5.0 ppb
- Trace Impurities - Silver (Ag):≤5.0 ppb
- Trace Impurities - Sodium (Na):≤5.0 ppb
- Trace Impurities - Strontium (Sr):≤5.0 ppb
- Trace Impurities - Tantalum (Ta):≤5.0 ppb
- Trace Impurities - Thallium (Tl):≤5.0 ppb
- Trace Impurities - Tin (Sn):≤5.0 ppb
- Trace Impurities - Titanium (Ti):≤5.0 ppb
- Trace Impurities - Vanadium (V):≤5.0 ppb
- Trace Impurities - Zinc (Zn):≤5.0 ppb
- Trace Impurities - Zirconium (Zr):≤5.0 ppb
- Particle Count at point of fill - 0.5 µm and greater:≤100 par/ml
- Particle Count at point of fill - 1.0 µm and greater:≤10 par/ml
Specification Test Results
About this item
:
ADR:8,II
:Management of Change (MOC) category = R
UN:2031
:GHS03
:GHS05
:GHS07
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349