Hexanes : acetone mixture 80:20 (v/v), ULTRA RESI-ANALYZED®, J.T.Baker®
Catalog # 9521-4B
Supplier: Avantor
CAS Number:
Specifications
- Pk:215 L
- Seq:5609
- Pk Info:1×215 L
Specifications
Specification Test Results
- Acetone - Assay (by GC, corrected for water):≥99.4 %
- Acetone - Color (APHA):≤10
- Acetone - Residue after Evaporation:≤1 ppm
- Acetone - Substances Reducing Permanganate:Passes Test
- Acetone - Titrable Acid (µeq/g):≤0.3
- Acetone - Titrable Base (µeq/g):≤0.60
- Water (H₂O)(by Karl Fischer titrn):≤0.5 %
- Acetone - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- Acetone - GC-ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (ng/L):≤10
- Hexanes - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- Hexanes - GC-ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (ng/L):≤10
- Hexanes - ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL):≤5
- Hexanes- Assay (Total Saturated C₆ Isomers) (by GC, corrected for water):≥99.5 %
- Hexanes- Assay (as n-hexane) (by GC):≥95 %
- Hexanes - Color (APHA):≤10
- Hexanes - Residue after Evaporation:≤1 ppm
- Hexanes - Substances Darkened by H₂SO₄:Passes Test
- Hexanes - Water (by KF, coulometric):≤0.05 %
Specification Test Results
About this item
:
ADR:3,II
:Management of Change (MOC) category = NI
Flash Pt:–23 °C
UN:1993
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient