Order Entry
Japan
ContactUsLinkComponent
N,N-Dimethylformamide ≥99.9% (by GC, corrected for water content) for HPLC, J.T.Baker®
Flammable
Irritant
Health
N,N-Dimethylformamide ≥99.9% (by GC, corrected for water content) for HPLC, J.T.Baker®
Catalog # 9220-98
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
Health
N,N-Dimethylformamide ≥99.9% (by GC, corrected for water content) for HPLC, J.T.Baker®
Catalog # 9220-98
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Stainless steel drum, CYCLE-TAINER
  • Pk:
    52 L
  • Seq:
    5382
  • Pk Info:
    1×52 L

Specifications

Specification Test Results

  • Appearance:
    Passes Test
  • Assay (by GC, corrected for water):
    ≥99.9 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤0.0005 %
  • Titrable Acid (meq/g):
    ≤0.0005
  • Titrable Base (meq/g):
    ≤0.003
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.03 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 270 nm:
    ≤0.75
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:
    ≤0.30
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 295 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 310 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm:
    ≤0.010

Specification Test Results

About this item

 :
 :73.09 g/mol
Density:0.95 g/cm³
UN:2265
ADR:3,III
 :Management of Change (MOC) category = N
Flash Pt:58.00 °C
 :GHS02
 :GHS07
 :GHS08
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284