

Petroleum spirit, 35…60 °C, AR® ACS, Macron Fine Chemicals™
Catalog # MACR4983-10
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth, PVC-coated
- Pk:4 L
- Seq:5162
- Pk Info:1×4 L
Specifications
Specification Test Results
- ACS - Acidity:Passes Test
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤0.001 %
- Appearance and Color:Passes Test
- ACS - Boiling Range(Lower):35 - 60 °C
- ACS - Boiling Range(Upper):35 - 60 °C
Specification Test Results
About this item
:
Boiling Pt:35…60 °C (1013 hPa)
EINECS:265-151-9
ADR:3,II
:Management of Change (MOC) category = N
UN:1268
:GHS02
:GHS08
Storage Temperature:Ambient
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849