

Calcium hydroxide ≥95.0%, powder, AR® ACS, Macron Fine Chemicals™
Catalog # MACR4195-05
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- パックタイプ:Case, Mall STAKMOR
- Pk:2,5 kg
- Seq:4076
- Pk Info:1×2,5 kg
Specifications
Specification Test Results
- ACS - Assay (Ca(OH)₂):≥95.0 %
- ACS - Assay CaCO₃:≤3 %
- ACS - Chloride (Cl):≤0.03 %
- ACS - Heavy Metals (as Pb):≤0.003 %
- ACS - Insoluble in HCl:≤0.03 %
- ACS - Iron (Fe):≤0.05 %
- ACS - Magnesium (Mg):≤0.5 %
- ACS - Potassium (K):≤0.05 %
- ACS - Sodium (Na):≤0.05 %
- ACS - Strontium (Sr):≤0.05 %
- ACS - Sulfur Compounds (as SO₄):≤0.1 %
Specification Test Results
About this item
:
:74.09 g/mol
Merck Index:13,01675
Melting Pt:550 °C
Density:2,24 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-137-3
UN:3262
ADR:8,III
:Management of Change (MOC) category = N
:GHS08
:GHS07
CAS Number:1305-62-0
Formula:Ca(OH)₂
MDL Number:MFCD00010901