Order Entry
Japan
ContactUsLinkComponent
Formaldehyde Test Strips, Dosatest
Formaldehyde Test Strips, Dosatest
Catalog # 85707.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Formaldehyde Test Strips, Dosatest
Catalog # 85707.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Triazol
  • Pk:
    100
  • Description:
    Formaldehyde test strips
  • Tests per kit:
    100
  • Range:
    0 • 10 • 20 • 40 • 60 • 100 • 200  mg/L HCHO

Specifications

About this item

Semi-quantitative test strips for the determination of formaldehyde.

  • Ideal for rapid and easy analysis directly on-site
  • No maintenance or calibration is required

Colour change: Beige to blue-violet

Delivery: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box