Order Entry
Japan
ContactUsLinkComponent
 
Potassium chloride 99.0-100.5% (by argentometric titration) USP, FCC, Macron Fine Chemicals™
 
undefined
Potassium chloride 99.0-100.5% (by argentometric titration) USP, FCC, Macron Fine Chemicals™

 

  • USP - Assay (KCl) (by Ag titrn):
    99.0 - 100.5 %
  • USP - Acidity or Alkalinity (no pink color then pink):
    Passes Test
  • USP - Bromide (Br)(USP):
    ≤0.1 %
  • USP - Calcium and Magnesium:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Iodide (I)(USP):
    ≤0.005 %
  • USP - Loss on Drying at 105°C:
    ≤1.0 %
  • USP - Sodium (Na):
    Passes Test
  • FCC - Assay (KCl) (by Ag titrn):
    99.0 - 100.5 %
  • FCC - Acidity or Alkalinity (no pink color then pink):
    Passes Test
  • FCC - Bromide and/or Iodide (FCC):
    Passes Test
  • FCC - Heavy Metals (as Pb)(FCC):
    ≤5 mg/kg
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Loss on Drying at 105°C:
    ≤1.0 %
  • FCC - Sodium (Na):
    Passes Test
  • Solubility (10 in 50):
    Passes Test

 

Frequently Bought Together