
Sodium chloride 99.0-100.5% (dry basis), granular FCC, USP, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2,16 g/cm³
:Management of Change (MOC) category = R
EINECS:231-598-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- USP - Assay (NaCl) (USP) (dried basis):99.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Acidity or Alkalinity:Passes Test
- USP - Aluminum (Al):≤0.2 ppm
- USP - Appearance of Solution:Passes Test
- USP - Arsenic (As):≤1 ppm
- USP - Barium (Ba):Passes Test
- USP - Nitrite (NO₂) (Absorbance Value):≤0.01
- USP - Magnesium and Alkaline Earth Metals (as Ca):≤100 ppm
- USP - Iodide (I):Passes Test
- USP - Bromide (Br):≤100 ppm
- USP - Iron (Fe):≤2 ppm
- USP - Loss on Drying:≤0.5 %
- USP - Phosphate (PO₄):≤25 ppm
- USP - Potassium (K):≤500 ppm
- USP - Ferrocyanide:Passes Test
- USP - Sulfate (SO₄):≤200 ppm
- FCC - Assay (as NaCl) (FCC) (ignited basis):99.0 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Arsenic (As):≤1 ppm
- FCC - Calcium and Magnesium (as Ca) (by EDTA titration)(FCC):≤0.35 %
- FCC - Heavy Metals (as Pb):≤2 ppm
- FCC - Iron (Fe):≤2 ppm
- FCC - Loss on Drying:≤0.5 %
- Appearance (White crystals or granules):Passes Test
Specification Test Results
Related Information
Frequently Bought Together