Order Entry
Japan
ContactUsLinkComponent
VWR®, Zinc Test Strips, Dosatest
VWR®, Zinc Test Strips, Dosatest
Catalog # 85700.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Zinc Test Strips, Dosatest
Catalog # 85700.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Dithizone
  • Pk:
    100
  • Description:
    Zinc test strips
  • Tests per kit:
    100
  • Range:
    0 • 2 • 5 • 10 • 25 • 50 • 100  mg/L Zn²⁺

Specifications

About this item

Semi-quantitative test strips for the determination of zinc.

  • Ideal for rapid and easy analysis of zinc directly on-site
  • No maintenance or calibration is required

Colour change: Orange to red

Delivery: Delivered as 100 test strips in a tube with reagents measuring vessel and instruction leaflet.
Caution: Strongly acid solutions (pH < 2) have to be buffered to a pH value of 2 to 6 using solid sodium acetate.