Order Entry
Japan
ContactUsLinkComponent
Phosphate test strips, Dosatest
Phosphate test strips, Dosatest
Catalog # 85704.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Phosphate test strips, Dosatest
Catalog # 85704.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Phosphomolybdenum blue
  • Pk:
    100
  • Description:
    Phosphate test strips
  • Tests per kit:
    100
  • Range:
    0 • 3 • 10 • 25 • 50 • 100  mg/L PO₄³

Specifications

About this item

Semi-quantitative test strips for the determination of phosphate.

  • Ideal for rapid and easy analysis of phosphate directly on-site
  • No maintenance or calibration is required

Colour change: White to blue-green

Delivery: Delivered as 100 test strips in a tube with instruction leaflet.