Order Entry
Japan
ContactUsLinkComponent
Copper test strips, Dosatest
Copper test strips, Dosatest
Catalog # 85696.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Copper test strips, Dosatest
Catalog # 85696.601
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Chemical method:
    Biquinoline
  • Pk:
    100
  • Description:
    Copper test strips
  • Tests per kit:
    100
  • Range:
    0 • 10 • 30 • 100 • 300  mg/L Cu⁺ ²⁺

Specifications

About this item

Semi-quantitative test strips for the determination of copper.

  • Ideal for rapid and easy analysis of copper directly on-site
  • No maintenance or calibration is required

Colour change: White to red-violet.

Delivery: Delivered as 100 test strips in a tube with instruction leaflet.
Caution: Strongly acid solutions (pH < 2) have to be buffered to a pH value of 2 to 6 using solid sodium acetate.