

Chloroform ≥99.8% (without stabilizer, GC), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9917-88
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤ 10
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤ 10
- Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water):≥ 99.8 %
- Chloride (Cl):≤ 10 ppm
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 2.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Titrable Acid (µeq/g):≤ 0.5
- Water (by KF, coulometric):≤ 0.05 %
Specification Test Results
About this item
:
:119.38 g/mol
ADR:6.1,III
:Management of Change (MOC) category = N
UN:1888
:GHS07
:GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826