Order Entry
Japan
ContactUsLinkComponent
Acetonitrile ≥99.8% (GC, on anhydrous basis), UltimAR®, Macron Fine Chemicals™
Flammable
Irritant
Acetonitrile ≥99.8% (GC, on anhydrous basis), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9911-88
CAS Number:  
Flammable
Irritant
Acetonitrile ≥99.8% (GC, on anhydrous basis), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9911-88
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤ 5
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤ 10
  • Assay (CH₃CN) (by GC, corrected for water):
    ≥ 99.8 %
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 1.0 ppm
  • Titrable Acid (µeq/g):
    ≤ 0.3
  • Titrable Base (µeq/g):
    ≤ 0.6
  • Water (by KF, coulometric):
    ≤ 0.05 %

Specification Test Results

About this item

 :
 :41.05 g/mol
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
UN:1648
ADR:3,II
 :Management of Change (MOC) category = N
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
REACH:01-2119471307-38
 :GHS02
 :GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878