Order Entry
Japan
ContactUsLinkComponent
Acetone ≥99.4% (GC, on anhydrous basis), UltimAR®, Macron Fine Chemicals™
Flammable
Irritant
Acetone ≥99.4% (GC, on anhydrous basis), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9910-88
CAS Number:  
Flammable
Irritant
Acetone ≥99.4% (GC, on anhydrous basis), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9910-88
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Assay ((CH₃)₂CO) (by GC, corrected for water):
    ≥ 99.4 %
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 1.0 ppm
  • Substances Reducing Permanganate:
    Passes Test
  • Titrable Acid (µeq/g):
    ≤ 0.3
  • Titrable Base (µeq/g):
    ≤ 0.6
  • Water (H₂O):
    ≤ 0.5 %
  • FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤ 5
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤ 10

Specification Test Results

About this item

 :
 :58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:≤20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
 :Management of Change (MOC) category = NI
REACH:01-2119471330-49
 :GHS02
 :GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765