Order Entry
Japan
ContactUsLinkComponent
Methanol ≥99.8% (by GC, corrected for water content), UltimAR®, Macron Fine Chemicals™
Flammable
Toxic
Health
Methanol ≥99.8% (by GC, corrected for water content), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9912-88
CAS Number:  
Flammable
Toxic
Health
Methanol ≥99.8% (by GC, corrected for water content), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9912-88
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤ 5
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤ 10
  • Assay (CH₃OH) (by GC, corrected for water):
    ≥ 99.8 %
  • Acetone:
    ≤ 0.001 %
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 1.0 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Titrable Acid (µeq/g):
    ≤ 0.3
  • Titrable Base (µeq/g):
    ≤ 0.10
  • Water (H₂O)(by coulometry):
    ≤ 0.08 %

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
 :Management of Change (MOC) category = NI
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595