- No. of blocks:1
- Weight:2,3 kg
- Description:Dry block heater
- Seq:01
- W×D×H:196×269×117 mm
- Temperature uniformity:±0,2 °C (within the block @ 37 °C)
- Pk:1
Multi-purpose, high wattage, constant temperature VWR® Standard Dry Block heaters are economical, versatile and compact.
- New design with smaller footprint
- Designed for multi-purpose use
- Exceptional temperature uniformity
- Holds interchangeable modular heating blocks
- Heats to 150 °C (302 °F)
- Electrical: 230 V, 50/60 Hz, 210 W
Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a variety of other laboratory procedures.
Each model accepts separate interchangeable modular blocks, which accommodate various sample enclosures such as vials, microplates, PCR strips, and a wide variety of tubes. Each block contains a thermometer well, and features close tube-and-block contact for maximum heat retention.
All units offer two separate temperature adjustment knobs providing reproducible low temperature and high temperature control. Low range knob adjusts from ambient to 100 °C (212 °F) and high range knob adjusts from 75 to 150 °C (167 to 302 °F).
Operating Conditions: Units can be run in conditions from 18 to 33 °C (64 to 91 °F) 20 to 80% relative humidity, non-condensing.