Order Entry
Japan
ContactUsLinkComponent
1,1,2,2-Tetrachloroethane ≥98% (by GC), J.T.Baker®
Toxic
Environment
1,1,2,2-Tetrachloroethane ≥98% (by GC), J.T.Baker®
Catalog # V398-08
Supplier:  Avantor
CAS Number:  
Toxic
Environment
1,1,2,2-Tetrachloroethane ≥98% (by GC), J.T.Baker®
Catalog # V398-08
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (CHCl₂CHCl₂) (by GC):
    ≥98 %
  • Appearance:
    Passes Test

Specification Test Results

About this item

 :
 :167.85 g/mol
Merck Index:13,09264
Boiling Pt:146.3 °C (1013 hPa)
Melting Pt:–43 °C
Density:1.595 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-197-8
UN:1702
ADR:6.1,II
 :Management of Change (MOC) category = N
 :GHS06
 :GHS09
CAS Number:79-34-5
Formula:Cl₂HCCHCl₂
MDL Number:MFCD00000848